3-Methylpentane 99+%

Supplier: Thermo Scientific Chemicals
Danger

12735-0010 12735-0500 12735-2500
TS12735-0010EA 414 USD
TS12735-0010 TS12735-0500 TS12735-2500
3-Methylpentane 99+%
3-Methylpentane
Formula: C₆H₁₄
MW: 86.18 g/mol
Boiling Pt: 64 °C (760 mmHg)
Melting Pt: –118 °C
Density: 0.66 g/cm³
Flash Pt: –32 °C
Storage Temperature: Ambient
MDL Number: MFCD00009342
CAS Number: 96-14-0
EINECS: 202-481-4
UN: 1208
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3755 to 1.3775 (20°C, 589 nm)
GC ≥99.0 %
Assay ≤8 % (Tetracyclin hydrochloride) On dry substance
Water ≤2.0 %
Sulfated ash ≤0.5 %
Identification Passes test
pH 2.3 to 3.3 (1 % in water CO2 free)
Specific optical rotation -235° to -250° (20°C, 589 nm) (c=0.25, water) (on dry substance)
Absorbance ≤0.40 (460 nm)
Heavy metals (as Pb) ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR