3-Methyl-2-cyclohexen-1-one 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methyl-2-cyclohexenone, 3-Methylcyclohex-2-enone

12666-0010 12666-0100 12666-0500
TS12666-0010EA 25 USD
TS12666-0010 TS12666-0100 TS12666-0500
3-Methyl-2-cyclohexen-1-one 98%
3-Methyl-2-cyclohexen-1-one
Formula: C₇H₁₀O
MW: 110.16 g/mol
Boiling Pt: 199…200 °C (760 mmHg)
Melting Pt: –21 °C
Density: 0.97 g/cm³
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00001581
CAS Number: 1193-18-6
EINECS: 214-769-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4935 to 1.4955 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR