2-Methylcyclohexanone 98%

Supplier: Thermo Scientific Chemicals
Warning

12661-0010 12661-0500 12661-2500
TS12661-0010EA 505 USD
TS12661-0010 TS12661-0500 TS12661-2500
2-Methylcyclohexanone 98%
2-Methylcyclohexanone
Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 162…163 °C (760 mmHg)
Melting Pt: –14 °C
Density: 0.924 g/cm³
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00001635
CAS Number: 583-60-8
EINECS: 209-513-6
UN: 2297
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4460 to 1.4500 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR