4-Methoxybenzenesulphonyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methoxybenzene-1-sulphonyl chloride

12573-0250 12573-1000
TS12573-0250EA 66 USD
TS12573-0250 TS12573-1000
4-Methoxybenzenesulphonyl chloride 99%
4-Methoxybenzenesulphonyl chloride
Formula: C₇H₇ClO₃S
MW: 206.65 g/mol
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 40…42 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007446
CAS Number: 98-68-0
EINECS: 202-692-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to beige Crystalline solid
Infrared spectrum Conforms
Melting point 36 to 41 °C
GC ≥98.5 %
Uniformity coefficient ≤2.00
Moisture holding capacity 60 to 70 % (H)
Total volume capacity ≥2.60 eq/l (H)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR