Methoxyacetyl chloride 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
12570-0100
12570-0500
12570-2500
TS12570-0100EA
77
USD
TS12570-0100
TS12570-0500
TS12570-2500
Methoxyacetyl chloride 97% stabilized
Methoxyacetyl chloride
Formula:
C₃H₅ClO₂ MW: 108.52 g/mol Boiling Pt: 112…113 °C (760 mmHg) Density: 1.187 g/cm³ Flash Pt: 28 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000728 CAS Number: 38870-89-2 EINECS: 254-169-2 UN: 2920 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4190 to 1.4240 (20°C, 589 nm) |
Stabilizer | 0.3 % MgO |
Titration RCOCl | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...