Bis(2-mercaptoethyl)sulfide 90%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,2'-Thiodiethanethiol, 2-Mercaptoethyl Sulphide

12549-0100 12549-0500
TS12549-0100EA 86 USD
TS12549-0100 TS12549-0500
Bis(2-mercaptoethyl)sulfide 90%
Bis(2-mercaptoethyl)sulfide
Formula: C₄H₁₀S₃
MW: 154.30 g/mol
Boiling Pt: 135…136 °C (10 mmHg)
Melting Pt: –11 °C
Density: 1.183 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00004893
CAS Number: 3570-55-6
UN: 3334
ADR: 9 III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5900 to 1.5980 (20°C, 589 nm)
GC ≥88 %
Non-volatile matter ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR