Ethyl-2-oxocyclopentaneacetate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Oxocyclopentaneacetic Acid Ethyl Ester, Ethyl 2-oxocyclopentylacetate, Ethyl 2-(2-Oxocyclopentyl)acetate, 2-(2-Oxocyclopentyl)acetic Acid Ethyl Ester, Ethyl 2-(2-oxocyclopentyl)acetate

12304-0010 12304-0050
TS12304-0010EA 77 USD
TS12304-0010 TS12304-0050
Ethyl-2-oxocyclopentaneacetate 95%
Ethyl-2-oxocyclopentaneacetate
Formula: C₉H₁₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00044715
CAS Number: 20826-94-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94 %
Specific gravity 0.859 to 0.864
Specific optical rotation -15° to -21° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR