Isophorone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5,5-Trimethyl-2-cyclohexen-1-one, 3,5,5-Trimethylcyclohex-2-enone

12264-0010 12264-0025 12264-5000
TS12264-0010EA 45 USD
TS12264-0010 TS12264-0025 TS12264-5000
Isophorone 97%
Isophorone
Formula: C₉H₁₄O
MW: 138.21 g/mol
Boiling Pt: 213…214 °C (760 mmHg)
Melting Pt: –8 °C
Density: 0.92 g/cm³
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00001584
CAS Number: 78-59-1
EINECS: 201-126-0
Merck Index: 13,05215

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4800 (20°C, 589 nm)
GC ≥96.0 %
NMR consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR