4-Hydroxy-3-methoxybenzylamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Vanillylamine HCl, 4-(Aminomethyl)-2-methoxyphenol hydrochloride

12137-0010 12137-0050
TS12137-0010EA 36 USD
TS12137-0010 TS12137-0050
4-Hydroxy-3-methoxybenzylamine hydrochloride 98%
4-Hydroxy-3-methoxybenzylamine hydrochloride
Formula: C₈H₁₂ClNO₂
MW: 189.64 g/mol
Melting Pt: 219…221 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012864
CAS Number: 7149-10-2
EINECS: 230-468-3

Order Now

Specification Test Results

Appearance Off-white Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR