6-Methyl-2-thiouracil 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methiocil, 2-Mercapto-6-methylpyrimidin-4(1H)-one, Methylthiouracil, MZU, Basethyrin, 4-Hydroxy-2-mercapto 6-methylpyrimidine, Thiothymin

12130-0025 12130-5000
TS12130-0025EA 296 USD
TS12130-0025 TS12130-5000
6-Methyl-2-thiouracil 98%
6-Methyl-2-thiouracil
Formula: C₅H₆N₂OS
MW: 142.18 g/mol
Melting Pt: 330 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006040
CAS Number: 56-04-2
EINECS: 200-252-3
Merck Index: 12,06210

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals and/or chunks
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR