4-Hydroxycoumarin 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Hydroxy-2-benzopyrone

12110-0050 12110-1000 12110-5000
TS12110-0050EA 23 USD
TS12110-0050 TS12110-1000 TS12110-5000
4-Hydroxycoumarin 98%
4-Hydroxycoumarin
Formula: C₉H₆O₃
MW: 162.14 g/mol
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 210…215 °C
MDL Number: MFCD00006856
CAS Number: 1076-38-6
EINECS: 214-060-2

Order Now

Specification Test Results

Appearance White to yellow to beige Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 210 to 217 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR