Ethyl-2-cyclohexanoneacetate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: ethyl 2-(2-oxocyclohexyl)acetate, (+/-)-2-(ethoxycarbonylmethyl)cyclohexanone, (2-oxo-cyclohexyl)-acetic acid ethyl ester

11832-0050 11832-0250
TS11832-0050EA 67 USD
TS11832-0050 TS11832-0250
Ethyl-2-cyclohexanoneacetate 97%
Ethyl-2-cyclohexanoneacetate
Formula: C₁₀H₁₆O₃
MW: 184.24 g/mol
Density: 1.02 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001638
CAS Number: 24731-17-7

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4570 to 1.4590 (20°C, 589 nm)
GC ≥96.0 %
SO4 and SO3 (as SO4) ≤0.1 %
Insoluble matter ≤0.005 %
Nitrogen compounds (as N) ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR