Ethyl-3-chloropropionate 98%

Supplier: Thermo Scientific Chemicals
Warning

11826-0250 11826-1000
TS11826-0250EA 45 USD
TS11826-0250 TS11826-1000
Ethyl-3-chloropropionate 98%
Ethyl 3-chloropropionate
Formula: C₅H₉ClO₂
MW: 136.58 g/mol
Boiling Pt: 162…163 °C (760 mmHg)
Density: 1.003 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00000989
CAS Number: 623-71-2
EINECS: 210-809-2
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4260 (20°C, 589 nm)
GC ≥97.5 %
Phosphate (PO4) ≤10 ppm
Calcium (Ca) ≤100 ppm
Iron (Fe) ≤10 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤100 ppm
Heavy metals (as Pb) ≤5 ppm (by ICP-OES)
Insoluble matter ≤0.01 %
Loss on ignition ≤0.5 %
Nitrogen compounds (as N) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR