Dithiooxamide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethanebis(thioamide), Rubeanic acid

11755-0050 11755-0250 11755-1000
TS11755-0050EA 103 USD
TS11755-0050 TS11755-0250 TS11755-1000
Dithiooxamide 98%
Dithiooxamide
Formula: C₂H₄N₂S₂
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004941
CAS Number: 79-40-3

Order Now

Specification Test Results

Appearance Orange to red to brown Powder
Infrared spectrum Conforms
Sulfated ash ≤2 %
Elemental Analysis 19.5 to 20.5 % (C)
Barium (Ba) ≤0.005 %
Calcium (Ca) ≤0.05 %
Iron (Fe) ≤0.01 %
Manganese (Mn) ≤5 ppm
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.5 %
Strontium (Sr) ≤0.005 %
Heavy metals (as Pb) ≤0.003 %
Insoluble matter ≤0.02 % (in dilute HCl)
Loss on ignition ≤2.0 %
Water-soluble substances ≤0.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR