Dimedone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 5,5-Dimethyl-1,3-cyclohexanedione, 5,5-Dimethylcyclohexane-1,3-dione

11615-0025 11615-0250 11615-1000 11615-5000
TS11615-0025EA 1275 USD
TS11615-0025 TS11615-0250 TS11615-1000 TS11615-5000
Dimedone 99%
Dimedone
Formula: C₈H₁₂O₂
MW: 140.18 g/mol
Melting Pt: 147…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00001588
CAS Number: 126-81-8
EINECS: 204-804-4
Merck Index: 13,03268

Order Now

Specification Test Results

Appearance White to yellow to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 146 to 150 °C
HPLC ≥98.5 %
Chloride (Cl) ≤5 ppm
Phosphate (PO4) ≤10 ppm
Sulfate (SO4) ≤5 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤2 ppm
KMnO4-reducing substance Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR