Diethylbenzene (mixture of isomers) mixture of o-, m- and p-isomers 95% (sum of isomers)

Supplier: Thermo Scientific Chemicals
Danger

11416-0010 11416-0025 11416-0100
TS11416-0010EA 66 USD
TS11416-0010 TS11416-0025 TS11416-0100
Diethylbenzene (mixture of isomers) mixture of o-, m- and p-isomers 95% (sum of isomers)
Diethylbenzene (mixture of isomers)
Formula: C₁₀H₁₄
Boiling Pt: 180…182 °C (760 mmHg)
Melting Pt: –73 °C
Density: 0.869 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00792900
CAS Number: 25340-17-4
EINECS: 246-874-9
UN: 2049
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.4960 (20°C, 589 nm)
GC ≥95.0 % (sum of o-, m-, p-isomers)
Titration with HCl 0.37 to 0.43 M
Bromide (Br) ≤500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR