4,7-Dichloroquinoline 98%

Supplier: Thermo Scientific Chemicals
Warning

11375-0050 11375-0250 11375-1000
TS11375-0050EA 22 USD
TS11375-0050 TS11375-0250 TS11375-1000
4,7-Dichloroquinoline 98%
4,7-Dichloroquinoline
Formula: C₉H₅Cl₂N
MW: 198.05 g/mol
Boiling Pt: 148 °C (760 mmHg)
Melting Pt: 84…86 °C
Flash Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00006774
CAS Number: 86-98-6
EINECS: 201-714-7

Order Now

Specification Test Results

Appearance White to yellow to brown Powder or crystals or crystalline solid
Infrared spectrum Conforms
Melting point 84 to 88 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR