3,6-Dichloropyridazine 97%

Supplier: Thermo Scientific Chemicals
Danger

11371-1000 11371-5000
TS11371-1000EA 108 USD
TS11371-1000 TS11371-5000
3,6-Dichloropyridazine 97%
3,6-Dichloropyridazine
Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Boiling Pt: 89…91 °C (0.2 mmHg)
Melting Pt: 65…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00006466
CAS Number: 141-30-0
EINECS: 205-478-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 65 to 70 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR