2,6-Dichloropurine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,6-Dichloro-9H-purine
11370-0010
11370-0050
TS11370-0010EA
139
USD
TS11370-0010
TS11370-0050
2,6-Dichloropurine 97%
2,6-Dichloropurine
Formula:
C₅H₂Cl₂N₄ MW: 189 g/mol Melting Pt: 184…186 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00077725 CAS Number: 5451-40-1 EINECS: 226-681-6 |
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...