1,4-Dichloro-2-butyne 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,4-Dichlorobut-2-yne

11328-0250 11328-1000
TS11328-0250EA 96 USD
TS11328-0250 TS11328-1000
1,4-Dichloro-2-butyne 99%
1,4-Dichloro-2-butyne
Formula: C₄H₄Cl₂
MW: 122.98 g/mol
Boiling Pt: 165…168 °C (760 mmHg)
Density: 1.25 g/cm³
Flash Pt: 160 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000981
CAS Number: 821-10-3
EINECS: 212-474-8
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5030 to 1.5060 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR