1,4-Dichlorobenzene 99+%

Supplier: Thermo Scientific Chemicals
Warning

11319-0010 11319-0025
TS11319-0010EA 58 USD
TS11319-0010 TS11319-0025
1,4-Dichlorobenzene 99+%
1,4-Dichlorobenzene
Formula: C₆H₄Cl₂
Boiling Pt: 174 °C (760 mmHg)
Melting Pt: 52…56 °C
Density: 1.24 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00000604
CAS Number: 106-46-7
EINECS: 203-400-5
UN: 3077
ADR: 9,III
Merck Index: 13,03082

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
Melting point 52 to 55 °C
GC ≥99.0 %
UV Lambda max 528 nm (in Methanol)
Fluorescence max 547 nm (in Methanol)
Optimum lasing 563 nm (in Methanol)
Suitability (for use as a laser dye) Passes test
Molar absorption coefficient 104000 l/(mole.cm) (Lambda max)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR