3,5-Dichloroaniline 98%

Supplier: Thermo Scientific Chemicals
Danger

11312-1000 11312-5000
TS11312-1000EA 61 USD
TS11312-1000 TS11312-5000
3,5-Dichloroaniline 98%
3,5-Dichloroaniline
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 260 °C (741 mmHg)
Melting Pt: 49…53 °C
Density: 1.58 g/cm³
Flash Pt: 133 °C
Storage Temperature: Ambient
MDL Number: MFCD00007774
CAS Number: 626-43-7
EINECS: 210-948-9
UN: 3442
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to brown Crystals (May darken on storage)
Infrared spectrum Conforms
Melting point 49 to 53 °C
GC ≥97.5 %
Fluorescence max 565 nm (in Methanol)
Optimum lasing 588 nm (in Methanol)
Molar absorption coefficient 106000 l/(mol.cm) (at Lambda max)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR