3,4-Dichloroaniline 98%

Supplier: Thermo Scientific Chemicals
Danger

11311-0010 11311-0050 11311-2500
TS11311-0010EA 218 USD
TS11311-0010 TS11311-0050 TS11311-2500
3,4-Dichloroaniline 98%
3,4-Dichloroaniline
Formula: C₆H₅Cl₂N
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 69…73 °C
Flash Pt: 166 °C
Storage Temperature: Ambient
MDL Number: MFCD00007768
CAS Number: 95-76-1
EINECS: 202-448-4
UN: 3442
ADR: 6.1,II
Merck Index: 13,03079

Order Now

Specification Test Results

Appearance White to grey to brown Crystals or flakes or crystalline mass and/or chunks (May darken on storage)
Infrared spectrum Conforms
Melting point 70 to 74 °C
GC ≥97.5 %
Specific optical rotation On anhydrous basis
Titration with HClO4 97.5 to 102.5 % (On anhydrous basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR