1,8-Dibromooctane 98%

Supplier: Thermo Scientific Chemicals

11288-0100 11288-0500 11288-2500
TS11288-0100EA 35 USD
TS11288-0100 TS11288-0500 TS11288-2500
1,8-Dibromooctane 98%
1,8-Dibromooctane
Formula: BrH₂C(CH₂)₆CH₂Br
Boiling Pt: 270…272 °C (760 mmHg)
Melting Pt: 15…16 °C
Density: 1.46 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000277
CAS Number: 4549-32-0
EINECS: 224-912-5

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4981 to 1.5001 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR