1,9-Dibromononane 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Nonamethylene dibromide

11287-0050 11287-0250
TS11287-0050EA 34 USD
TS11287-0050 TS11287-0250
1,9-Dibromononane 97%
1,9-Dibromononane
Formula: BrH₂C(CH₂)₇CH₂Br
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: –10 °C
Density: 1.40 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000279
CAS Number: 4549-33-1
EINECS: 224-913-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4950 to 1.4980 (20°C, 589 nm)
GC ≥96.0 %
Specific optical rotation +39° to +69° (20°C, 589 nm) (C=2, Chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR