Dibromomethane 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methylenbromide

11283-5000 11283-0010 11283-0025 11283-1000
TS11283-5000EA 89 USD
TS11283-5000 TS11283-0010 TS11283-0025 TS11283-1000
Dibromomethane 99%
Dibromomethane
Formula: Br₂CH₂
MW: 173.83 g/mol
Boiling Pt: 96…98 °C (760 mmHg)
Melting Pt: –52 °C
Density: 2.47 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000168
CAS Number: 74-95-3
EINECS: 200-824-2
UN: 2664
ADR: 6.1,III
Merck Index: 12,06138

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5400 to 1.5420 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR