1,6-Dibromohexane 98%

Supplier: Thermo Scientific Chemicals
Warning

11281-0025 11281-1000 11281-5000
TS11281-0025EA 490 USD
TS11281-0025 TS11281-1000 TS11281-5000
1,6-Dibromohexane 98%
1,6-Dibromohexane
Formula: BrH₂C(CH₂)₄CH₂Br
Boiling Pt: 243 °C (760 mmHg)
Melting Pt: –2 °C
Density: 1.58 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000272
CAS Number: 629-03-8
EINECS: 211-067-2
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5056 to 1.5076 (20°C, 589 nm)
GC ≥97.5 %
Specific optical rotation +29° to +33° (20°C, 589 nm) (c=1, toluene)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR