1,10-Dibromodecane 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,10-Dibromodecan, Decane, 1,10-dibromo-

11278-0250 11278-1000 11278-5000
TS11278-0250EA 61 USD
TS11278-0250 TS11278-1000 TS11278-5000
1,10-Dibromodecane 97%
1,10-Dibromodecane
Formula: BrH₂C(CH₂)₈CH₂Br
MW: 300.08 g/mol
Boiling Pt: 160 °C (15 mmHg)
Melting Pt: 25…30 °C
Density: 1.34 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000222
CAS Number: 4101-68-2
EINECS: 223-871-0

Order Now

Specification Test Results

Appearance White to light brown Fused solid or liquid
Infrared spectrum Conforms
Melting point 25 to 30 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR