Dibenzoylmethane 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,3-Diphenyl-1,3-propanedione, 1,3-Diphenylpropane-1,3-dione

11260-0100 11260-0250 11260-1000 11260-5000
TS11260-0100EA 28 USD
TS11260-0100 TS11260-0250 TS11260-1000 TS11260-5000
Dibenzoylmethane 98%
Dibenzoylmethane
Formula: (C₆H₅)C(O)CH₂C(O)(C₆H₅)
MW: 224.26 g/mol
Boiling Pt: 219…221 °C (18 mmHg)
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00003085
CAS Number: 120-46-7
EINECS: 204-398-9
Merck Index: 13,03033

Order Now

Specification Test Results

Appearance Yellow Crystals
Infrared spectrum Conforms
Melting point 75 to 79 °C
Sulfated ash ≤0.1 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR