(Bromomethyl)cyclohexane 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11135-0050
11135-0250
11135-1000
TS11135-0050EA
28
USD
TS11135-0050
TS11135-0250
TS11135-1000
(Bromomethyl)cyclohexane 96%
(Bromomethyl)cyclohexane
Formula:
C₇H₁₃Br Boiling Pt: 76…77 °C (26 mmHg) Density: 1.28 g/cm³ Flash Pt: 57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001509 CAS Number: 2550-36-9 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Clear light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4910 to 1.4930 (20°C, 589 nm) |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...