2-Cyclohexen-1-one 97%

Supplier: Thermo Scientific Chemicals
Danger

11124-5000 11124-0100 11124-0250 11124-1000
TS11124-5000EA 684 USD
TS11124-5000 TS11124-0100 TS11124-0250 TS11124-1000
2-Cyclohexen-1-one 97%
2-Cyclohexen-1-one
Formula: C₆H₈O
MW: 96.13 g/mol
Boiling Pt: 168 °C (760 mmHg)
Melting Pt: –53 °C
Density: 0.993 g/cm³
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00001577
CAS Number: 930-68-7
EINECS: 213-223-5
UN: 2929
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4875 to 1.4895 (20°C, 589 nm)
GC ≥96.0 %
Absorption Ratio P-15/P+15 1.01 to 1.16
BSC Certification Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR