1,3-Cyclohexanedione 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cyclohexane-1,3-dione
11117-1000
11117-5000
TS11117-1000EA
65
USD
TS11117-1000
TS11117-5000
1,3-Cyclohexanedione 97%
1,3-Cyclohexanedione
Formula:
C₆H₈O₂ MW: 112.13 g/mol Melting Pt: >101 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001585 CAS Number: 504-02-9 EINECS: 207-980-0 Merck Index: 13,03200 |
Specification Test Results
Appearance | Light yellow to beige Powder |
Infrared spectrum | Conforms |
Melting point | ≥101 °C |
Titration Argentometric | ≤1 % Sodium chloride |
Water | ≤0.5 % |
Titration with NaOH | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...