1,2-Cyclohexanedione 98%

Supplier: Thermo Scientific Chemicals

11116-0100 11116-0500 11116-2500
TS11116-0100EA 173 USD
TS11116-0100 TS11116-0500 TS11116-2500
1,2-Cyclohexanedione 98%
1,2-Cyclohexanedione
Formula: C₆H₈O₂
Boiling Pt: 193…195 °C (760 mmHg)
Melting Pt: 35…38 °C
MDL Number: MFCD00001648
CAS Number: 765-87-7
EINECS: 212-155-3

Order Now

Specification Test Results

Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5180 to 1.5200 (20°C, 589 nm)
GC ≥97.5 %
Copper (Cu) ≤0.0005 %
Iron (Fe) ≤0.005 %
Magnesium (Mg) ≤0.01 %
Manganese (Mn) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR