trans-Cinnamaldehyde 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11035-0010
11035-0050
11035-0500
11035-5000
TS11035-0010EA
62
USD
TS11035-0010
TS11035-0050
TS11035-0500
TS11035-5000
trans-Cinnamaldehyde 99%
trans-Cinnamaldehyde
Formula:
C₉H₈O MW: 132.16 g/mol Boiling Pt: 252 Melting Pt: -8 Density: 1.05 g/cm³ Flash Pt: 71 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007000 CAS Number: 14371-10-9 Merck Index: 12,02319 |
Specification Test Results
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6210 to 1.6230 (20°C, 589 nm) |
GC | ≥98.5 % |
Stabilizer | 2,6-di-tert.-Butyl-p-cresol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...