Chrysin 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: 5,7-Dihydroxy-2-phenyl-4H-chromen-4-one, 5,7-Dihydroxyflavone, 5,7-Dihydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 5,7-Dihydroxy-2-phenyl-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-phenyl-4-benzopyrone, 5,7-Dihydroxy-2-phenylchromen-4-one

11032-0050 11032-0250 11032-1000
TS11032-0050EA 46 USD
TS11032-0050 TS11032-0250 TS11032-1000
Chrysin 99+%
Chrysin
Formula: C₁₅H₁₀O₄
MW: 254.24 g/mol
Boiling Pt: 429
Melting Pt: 284…290 °C
Storage Temperature: Ambient
MDL Number: MFCD00006834
CAS Number: 480-40-0
EINECS: 207-549-7

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 284 to 290 °C
Loss on drying ≤1.0 %
HPLC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR