4-Chloroquinoline 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11005-0010
11005-0050
TS11005-0010EA
111
USD
TS11005-0010
TS11005-0050
4-Chloroquinoline 99%
4-Chloroquinoline
Formula:
C₉H₆ClN MW: 163.61 g/mol Boiling Pt: 260…261 °C (760 mmHg) Melting Pt: 29…32 °C Density: 1.25 g/cm³ Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006773 CAS Number: 611-35-8 |
Specification Test Results
Appearance | Light yellow Low melting solid |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...