3-Chloropyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

11002-0250 11002-1000
TS11002-0250EA 57 USD
TS11002-0250 TS11002-1000
3-Chloropyridine 99%
3-Chloropyridine
Formula: C₅H₄ClN
MW: 113.55 g/mol
Boiling Pt: 147…150 °C (760 mmHg)
Density: 1.19 g/cm³
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00006375
CAS Number: 626-60-8
EINECS: 210-955-7

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5310 to 1.5340 (20°C, 589 nm)
GC ≥98.5 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 Passes test
Carbonyl compounds ≤0.001 % (each of acetone, formaldehyde, acetaldehyde)
Titrable acid ≤0.0003 meq/g
Titrable base ≤0.0002 meq/g
KMnO4-reducing substance Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR