1-Chloropropane 99%

Supplier: Thermo Scientific Chemicals
Danger

10995-0050 10995-1000 10995-5000
TS10995-0050EA 43 USD
TS10995-0050 TS10995-1000 TS10995-5000
1-Chloropropane 99%
1-Chloropropane
Formula: C₃H₇Cl
Boiling Pt: 46…47 °C (760 mmHg)
Melting Pt: –123 °C
Density: 0.892 g/cm³
Flash Pt: –18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000995
CAS Number: 540-54-5
EINECS: 208-749-7
UN: 1278
ADR: 3,II
Merck Index: 12,08032

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3870 to 1.3900 (20°C, 589 nm)
GC ≥98.5 %
Iron (Fe) ≤50 ppm
Mercurous Mercury (as Hg) ≤0.15 %
Residue after reduction ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR