2-Chloro-2-methylpropane 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: tert-Butyl chloride

10950-0025 10950-0250 10950-5000
TS10950-0025EA 233 USD
TS10950-0025 TS10950-0250 TS10950-5000
2-Chloro-2-methylpropane 99%
2-Chloro-2-methylpropane
Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 51 °C (1013 hPa)
Melting Pt: –25 °C
Density: 0.85 g/cm³
Flash Pt: –27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000816
CAS Number: 507-20-0
EINECS: 208-066-4
UN: 1127
ADR: 3,II
Merck Index: 13,01561

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3838 to 1.3858 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤15 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR