1-(Chloromethyl)naphthalene 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
10945-0050
10945-1000
10945-5000
TS10945-0050EA
26
USD
TS10945-0050
TS10945-1000
TS10945-5000
1-(Chloromethyl)naphthalene 95%
1-(Chloromethyl)naphthalene
Formula:
C₁₁H₉Cl MW: 176.65 g/mol Boiling Pt: 291 °C (1013 hPa) Melting Pt: 31…31 °C Density: 1.17 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004042 CAS Number: 86-52-2 EINECS: 201-678-2 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | Clear yellow to brown Liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.6350 to 1.6390 (20°C, 589 nm) |
HPLC | Remainder 2-chloromethylnaphtalene |
Acid value | ≤35 mg KOH/g |
Peroxide value | ≤4 meq O2/kg |
UV | at 280 nm A: ≤0.1 |
Residue after ign. (sulfated) | ≤0.3 % |
Specific optical rotation | (after 24 h) |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...