4-Chlorobutyryl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Chlorobutanoyl Chloride
10890-0010
10890-2500
TS10890-0010EA
118
USD
TS10890-0010
TS10890-2500
4-Chlorobutyryl chloride 98%
4-Chlorobutyryl chloride
Formula:
C₄H₆Cl₂O Boiling Pt: 173…174 °C (760 mmHg) Melting Pt: –47 °C Density: 1.25 g/cm³ Flash Pt: 85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000754 CAS Number: 4635-59-0 EINECS: 225-059-1 UN: 2922 ADR: 8,I |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4605 to 1.4625 (20°C, 589 nm) |
GC | ≥99.0 % |
Solubility | (in water) Passes test |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Carbonyl compounds | ≤0.02 % 2-Butanone |
Titrable acid | ≤0.0005 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...