trans-3-Chloroacrylic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

10855-0010 10855-0050
TS10855-0010EA 96 USD
TS10855-0010 TS10855-0050
trans-3-Chloroacrylic acid 99%
trans-3-Chloroacrylic acid
Formula: C₃H₃ClO₂
MW: 106.51 g/mol
Melting Pt: 82…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00064237
CAS Number: 2345-61-1
EINECS: 219-070-0
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance Beige to grey Powder or flakes
Infrared spectrum Conforms
Melting point 82 to 85 °C
Titration with NaOH ≥98.5 %
Clarity of alcohol solution Passes test
Ammonium (NH4) Passes test
Iron (Fe) ≤5 ppm
Heavy metals (ICP-OES) ≤5 ppm
Sulfur compounds (as SO4) ≤0.005 %
Titrable free acid ≤0.25 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR