Dibutyl sulphide 99%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dibutylsulphide, Butyl sulphide, n-Butyl sulphide
10801-5000
10801-0025
10801-1000
TS10801-5000EA
221
USD
TS10801-5000
TS10801-0025
TS10801-1000
Dibutyl sulphide 99%, pure
Dibutyl sulphide
Formula:
C₈H₁₈S MW: 146.3 g/mol Boiling Pt: 189 Melting Pt: -80 Density: 0.84 g/cm³ Flash Pt: 62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009468 CAS Number: 544-40-1 EINECS: 208-870-5 UN: 3334 ADR: 9 III Merck Index: 12,01626 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4520 to 1.4540 (20°C, 589 nm) |
GC | ≥98.5 % |
Sulfated ash | ≤0.05 % |
Solubility | (3 M in water) Clear colorless |
UV | (3 M in water) |
Sulfate (SO4) | ≤50 ppm |
Iron (Fe) | ≤1 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...