4-tert-Butylcyclohexanone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
10791-0250
10791-1000
10791-5000
TS10791-0250EA
28
USD
TS10791-0250
TS10791-1000
TS10791-5000
4-tert-Butylcyclohexanone 99%
4-tert-Butylcyclohexanone
Formula:
C₁₀H₁₈O Boiling Pt: 113…116 °C (20 mmHg) Melting Pt: 48…51 °C Flash Pt: 96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001642 CAS Number: 98-53-3 EINECS: 202-678-5 |
Specification Test Results
Appearance | White to light yellow Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 46 to 51 °C |
GC | ≥98.5 % |
Water | ≤1.0 % (K.F.) |
Anticaking agent | 0.5 % (SiO2) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...