tert-Butylacetyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,3-Dimethylbutyryl chloride

10779-0100 10779-0500 10779-2500
TS10779-0100EA 40 USD
TS10779-0100 TS10779-0500 TS10779-2500
tert-Butylacetyl chloride 99%
tert-Butylacetyl chloride
Formula: C₆H₁₁ClO
MW: 134.61
Boiling Pt: 127…129 °C (760 mmHg)
Density: 0.96 g/cm³
Flash Pt: 21 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000737
CAS Number: 7065-46-5
EINECS: 230-356-4
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4210 to 1.4230 (20°C, 589 nm)
Titration RCOCl ≥98.5 %
Titration RCOOH ≤1 %
pH 3.8 to 7.6 (Ph. Eur.(c=1, about 55°C))
Arsenic (As) ≤1 ppm
Lead (Pb) ≤5 ppm
Heavy metals (as Pb) ≤50 ppm
Sulfur dioxide (SO2) ≤40 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR