3-Bromopyridine 99%

Supplier: Thermo Scientific Chemicals
Danger

10741-0250 10741-1000
TS10741-0250EA 86 USD
TS10741-0250 TS10741-1000
3-Bromopyridine 99%
3-Bromopyridine
Formula: C₅H₄BrN
MW: 158 g/mol
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: –27 °C
Density: 1.64 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00006373
CAS Number: 626-55-1
EINECS: 210-952-0
UN: 2929
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5680 to 1.5720 (20°C, 589 nm)
GC ≥98.5 %
UV at 280 nm A: ≤0.005
Color scale ≤10 APHA
Stabilizer 2 % Ethanol
Wavelength at 1 A.U. ≤205 nm
Residue after evaporation ≤2 ppm
Acidity/alkalinity ≤0.0002 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR