2-Bromopropene 99% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Propenyl bromide, α-Methylvinyl bromide, Isopropenyl bromide

10734-0100 10734-0500
TS10734-0100EA 88 USD
TS10734-0100 TS10734-0500
2-Bromopropene 99% stabilized
2-Bromopropene
Formula: C₃H₅Br
MW: 120.98 g/mol
Boiling Pt: 47…49 °C (760 mmHg)
Melting Pt: –125 °C
Density: 1.36 g/cm³
Flash Pt: 4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000140
CAS Number: 557-93-7
EINECS: 209-185-4
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4420 to 1.4460 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer Approx. 100 ppm BHT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR