2-Bromopropane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
10732-0010
10732-0025
10732-0100
10732-2500
TS10732-0010EA
194
USD
TS10732-0010
TS10732-0025
TS10732-0100
TS10732-2500
2-Bromopropane 99%
2-Bromopropane
Formula:
H₃CBrCHCH₃ Boiling Pt: 59 °C (760 mmHg) Melting Pt: –89 °C Density: 1.31 g/cm³ Flash Pt: 1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000147 CAS Number: 75-26-3 EINECS: 200-855-1 UN: 2344 ADR: 3,II Merck Index: 13,05230 |
Specification Test Results
Appearance | Clear colorless to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4250 to 1.4270 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...