(3-Bromopropyl)benzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-3-phenylpropane
10729-0050
10729-1000
10729-5000
TS10729-0050EA
33
USD
TS10729-0050
TS10729-1000
TS10729-5000
(3-Bromopropyl)benzene 98%
(3-Bromopropyl)benzene
Formula:
BrH₂CCH₂CH₂(C₆H₅) Boiling Pt: 219…220 °C (760 mmHg) Density: 1.31 g/cm³ Flash Pt: 101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000257 CAS Number: 637-59-2 EINECS: 211-294-7 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5450 to 1.5470 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...