1-Bromononane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Nonyl bromide

10720-0250 10720-1000 10720-5000
TS10720-0250EA 49 USD
TS10720-0250 TS10720-1000 TS10720-5000
1-Bromononane 97%
1-Bromononane
Formula: BrH₂C(CH₂)₇CH₃
Boiling Pt: 201 °C (760 mmHg)
Density: 1.08 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00000278
CAS Number: 693-58-3
EINECS: 211-755-2

Order Now

Specification Test Results

Appearance Clear colorless to golden Liquid
Infrared spectrum Conforms
Refractive index 1.4530 to 1.4550 (20°C, 589 nm)
GC ≥96.0 %
Absorption Ratio P-15/P+15 0.96 to 1.22
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR